HPLC Analysis of Terfenadine Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Terfenadine Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 40 °C
mobile phase [A] heptane; [B] ethanol; [C] diethylamine; (98:2:0.1, A:B:C)
flow rate 0.5 mL/min
sample 1 g/L in methanol
injection 3 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Anti-inflammatories, Antihistamines
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC