HPLC Analysis of Tetracycline Antibiotics on Ascentis® Phenyl
HPLC Analysis of Tetracycline Antibiotics on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 C
mobile phase [A] 0.1% ammonium acetate; [B] acetonitrile with 0.1% ammonium acetate; (80:20, A:B)
flow rate 1.0 ml/min
sample 50 µg/mL each in 80:20, water:methanol
injection 10 µl
detector UV, 353 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antibiotics, Antimicrobials