Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tetramisole Enantiomers and Related Compounds on Astec® CYCLOBOND® I 2000 DMP
HPLC Analysis of Tetramisole Enantiomers and Related Compounds on Astec® CYCLOBOND® I 2000 DMP
Conditions
column Astec CYCLOBOND I 2000 DMP 10 cm X 2.1 mm
column temp. 35 C
mobile phase (A) 100 mM ammonium acetate, pH 5 with acetic acid (B) acetonitrile, 90:10, v/v
flow rate 0.2 ml/min
pressure 360 psi
sample 100 ug/mL tetramisole and 50 ug/mL each cocaine and aminorex in water:methanol, 80:20, v/v
injection 2 µl
detector UV, 220 nm
Description
Analysis Note Cocaine is often found laced with levamisole (one of the tetramisole enantiomers) and aminorex is a metabolite of levamisole known to cause serious vascular damage in drug abusers. These compounds are readily separated using the Astec CYCLOBOND column - a derivatized cyclodextrin column only offered by Supelco. What is really interesting about this separation is that it shows the ability of chiral HPLC columns to separate enantiomers (here, dexamisole and levamisole) by chiral discrimination, as well as non-chiral discrimination in the same analysis. The compounds aminorex and cocaine are not enantiomers of each other or of tetramisole. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Categories Analytical Chromatography, Antihelminthic, Drugs of Abuse
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC