HPLC Analysis of Thalidomide Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Thalidomide Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 µL
mobile phase methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Anxiolytics, Sedatives, Pharmaceutical (small molecule), All Pharmaceutical Applications, Anxiolytics/Sedatives
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC