HPLC Analysis of the Radiocontrast Agent Iohexol on SeQuant® ZIC-HILIC
HPLC Analysis of the Radiocontrast Agent Iohexol on SeQuant® ZIC-HILIC
Conditions
column Sequant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase [A] 50 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate 1.5 mL/min
pressure 696 psi (48 bar)
column temp. 40 °C ambient
detector 254 nm (cell: 13 μL)
sample 100 ppm iohexol in mobile phase
injection: 5 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany