Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Thyroglobulin on a SRT® SEC-1000 Gel Filtration Column
HPLC Analysis of Thyroglobulin on a SRT® SEC-1000 Gel Filtration Column
Conditions
column SRT SEC-1000, 30 cm x 4.6 mm I.D., 5 µm particles, 1000 Ã… (Z777061)
column temp. Ambient
mobile phase 150 mM phosphate buffer, pH 7.0
flow rate 0.35 ml/min
sample 1. Thyroglobulin (670kD) 1 mg/mL; 2. Uracil (120D), 0.1 mg/mL
injection 3 µl
detector UV, 214 nm
Description
Analysis Note Thyroglobulin is a very large protein, with a molecular mass of 670 kDa. Thus,a large pore size gel filtration columns is required to prevent partial exclusion of not just the monomer, but also of the dimer and higher order aggregates. This was accomplished using the Sepax SRT SEC-1000 column, which has a nominal pore size of 1000 Angstrom. The nucleic acid base Uracil was added to the sample as a marker to indicate the inclusion volume since a small molecule like Uracil is not restricted from entering any pores in the column.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies