HPLC Analysis of Toluidine Isomers on Ascentis® Si (Silica), (80:20 Hexane:Ethyl Acetate)
HPLC Analysis of Toluidine Isomers on Ascentis® Si (Silica), (80:20 Hexane:Ethyl Acetate)
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 35 C
mobile phase [A] hexane; [B] ethyl acetate; (80:20, A:B)
flow rate 1.0 ml/min
sample 50 µg/mL in mobile phase
injection 10 µl
detector UV, 245 nm
Description
Categories Analytical Chromatography, Amines
Featured Industry Chemicals and Industrial Polymers