HPLC Analysis of Toluidine Isomers on Ascentis® Si (Hexane: 2-Propanol)
HPLC Analysis of Toluidine Isomers on Ascentis® Si (Hexane: 2-Propanol)
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 35 °C
mobile phase [A] hexane; [B] 2-propanol; (96:4, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL in mobile phase
injection 10 µL
detector UV, 245 nm
Description
Categories Analytical Chromatography, Amines
Featured Industry Chemicals and Industrial Polymers