HPLC Analysis of Tröger′s Base Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Tröger′s Base Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 °C
mobile phase [A] heptane; [B] IPA; [C] DEA; (90:10:0.1, A:B:C)
flow rate 0.5 mL/min
sample 2 mg/mL in mobile phase
injection 2 µL
detector UV, 254 nm
Description
Application Tröger′s base is an organic compound that exhibits chirality due to the presence of two bridgehead stereogenic nitrogen atoms in its structure.
Categories Analytical Chromatography, Heterocyclic compounds
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC