Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tröger′s Base Enantiomers on Astec® Cellulose DMP (LC/MS Mobile Phase)
HPLC Analysis of Tröger′s Base Enantiomers on Astec® Cellulose DMP (LC/MS Mobile Phase)
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 C
mobile phase [A] methanol; [B] ammonium formate; (100:0.1, A:B)
flow rate 0.5 ml/min
injection 2 µl
detector UV, 230 nm
Description
Application Tröger′s base is an organic compound that exhibits chirality due to the presence of two bridgehead stereogenic nitrogen atoms in its structure.
Categories Analytical Chromatography, Heterocyclic compounds
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC