HPLC Analysis of Triacylglycerols in Sacha Inchi Oil on Ascentis® Express C18
HPLC Analysis of Triacylglycerols in Sacha Inchi Oil on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
mobile phase [A] acetonitrile; [B] 2-propanol
gradient 0 min 0%; 50 min 70% B; 55 min 70% B; 56 min 0% B.
flow rate 1 ml/min
injection 2 µl
detector APCI/MS (positive), TIC
Description
Analysis Note An Ascentis Express C18 column packed with 2.7 μm Fused-Core particles was used to to perform this analysis.
Categories Analytical Chromatography, Triglycerides
Featured Industry Food and Beverages