HPLC Analysis of Tricyclic Antidepressants on Discovery® RP-AmideC16
HPLC Analysis of Tricyclic Antidepressants on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
mobile phase [A] water/acetonitrile (40:60 v/v); [B] 10 mM potassium phosphate
flow rate 2 ml/min
injection 10 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antidepressants
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC