HPLC Analysis of Tricyclic Antidepressants on Ascentis® Express C18
HPLC Analysis of Tricyclic Antidepressants on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 µm particles (53823-U)
column temp. 55 °C
mobile phase [A] 100 mM ammonium acetate (pH 7.0; titrated with ammonium hydroxide), [B] water, [C] methanol
flow rate 0.3 mL/min
injection 1 µL
detector Thermo LCQ Advantage; ESI(+), m/z 250-320
instrument Jasco X-LC
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Clinical, antidepressants, Antidepressants
Featured Industry Clinical
Pharmaceutical (small molecule)