HPLC Analysis of Tripeptide Gly-His-Lys on SeQuant ZIC-HILIC
HPLC Analysis of Tripeptide Gly-His-Lys on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm (1.50455.0001)
mobile phase [A] acetonitrile; [B] ammonium formate; (40:60, A:B)
flow rate 0.5 mL/min
injection 2 µL
detector 214 nm
Description
Categories Analytical Chromatography, Peptides