HPLC Analysis of Trypsin on TSKgel® Butyl-NPR
HPLC Analysis of Trypsin on TSKgel® Butyl-NPR
Conditions
column TSKgel Butyl-NPR, 3.5 cm x 4.6 mm I.D., 2.5 µm particles (814947)
mobile phase 2 M to 0 M ammonium sulfate in 20 mM Tris-HCI buffer, pH 7.5 in 10 min
flow rate 1 ml/min
injection commercial trypsin sample
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation