HPLC Analysis of Tryptophan Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
HPLC Analysis of Tryptophan Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] water; [B] methanol; [C] formic acid; (30:70:0.02, A:B:C)
flow rate 1 mL/min
sample 300 mug /mL in 30:70 water:methanol
injection 10 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC