HPLC Analysis of Tryptophan Hydroxylase Inhibitors on Ascentis® C18
HPLC Analysis of Tryptophan Hydroxylase Inhibitors on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 C
mobile phase [A] 40 mM formic acid, (pH ~2.5): [B] methanol; (65:35, A:B)
flow rate 0.7 ml/min
sample as indicated in 90:10, 10 mM formic acid:methanol
injection 10 µl
detector full scan MS; overlay of extracted ion chromatograms of individual (M+H)+ species
Description
Categories Analytical Chromatography, Clinical, proteins, Amino Acids
Featured Industry Clinical
Pharmaceutical (small molecule)