HPLC Analysis of Typical Parent Antipsychotic Drugs Using an Ascentis® Express C18 Column
HPLC Analysis of Typical Parent Antipsychotic Drugs Using an Ascentis® Express C18 Column
Conditions
column Ascentis Express C18, 10 cm  x 3.0 mm I.D., 1.9 µm particles (50819-U)
column temp. 35 °C
mobile phase [A] 0.1% formic acid in water, [B] 0.1 formic acid in methanol
gradient 25% B for 2 minutes followed by a linear gradient to 95% B for 10 minutes, followed by a hold at 95% B for 2 minutes.
flow rate 0.5 mL/min
pressure 5900 psi (407 bar)
sample 25 µg/mL each in 10% methanol
injection 2 µL
detector UV, 254 nm
Description
Analysis Note An application was developed on an Ascentis Express C18 column to separate typical parent antipsychotics.
Categories Analytical Chromatography, Antipsychotics