HPLC Analysis of Urea, Carbamate, Triazine, Aniline Pesticides on Discovery® C18
HPLC Analysis of Urea, Carbamate, Triazine, Aniline Pesticides on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 20 °C
mobile phase [A] acetonitrile; [B] water
flow rate 2 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Pesticides & Herbicides, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Environmental
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC