HPLC Analysis of Urea on SeQuant ZIC-HILIC
HPLC Analysis of Urea on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 µm, 200 Ã… (1.50451.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate 5 mM; (90:10, A:B)
flow rate 0.5 mL/min
injection 5 µL
detector 204 nm
Description
Categories Analytical Chromatography, Amines
Featured Industry Chemicals and Industrial Polymers
Life Science and Biopharma