Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Veterinary Antibiotics on Ascentis® C18
HPLC Analysis of Veterinary Antibiotics on Ascentis® C18
Conditions
column Ascentis C18, 5 cm x 2.1 mm I.D., 3 µm particles (581300-U)
column temp. 22 C
mobile phase [A] 50 mM ammonium acetate (pH 6.0): [B] methanol; (20:80, A:B)
flow rate 0.2 ml/min
sample as indicated in 50:50, water:methanol
injection 2 µl
detector ESI(-), full scan
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, antibiotics/antimicrobials, Antibiotics, Antimicrobials, Antibiotics
Featured Industry Clinical