HPLC Analysis of Vinblastine Sulfate on Chromolith® HighResolution RP-18 endcapped 150 x 4.6mm
HPLC Analysis of Vinblastine Sulfate on Chromolith® HighResolution RP-18 endcapped 150 x 4.6mm
Conditions
sample preparation Transfer USP Vincristine Sulfate RS  ( or Assay RS) to a suitable volumetric flask, and dissolve in the Standard solution.
column Chromolith® HighResolution RP-18 endcapped, 150 x 4.6 mm (Merck p/n 1.52023) (1.52023.0001)
column temp. ambient
mobile phase [A] Dimethylamine and water 14/986 (v/v). Adjusted with phosphoric acid to a pH of 7.5; [B] Acetonitrile and Methanol 20/80 (v/v); (38:62, A:B)
flow rate 2.0 mL/min
pressure 2030 psi (140 bar)
sample 0.4 mg/mL each of Vincristine sulfate  and Vinblastine Sulfate in water (Diluent: water)
injection 20 µL
detector cell: 11 µL
detector 262 nm
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany