HPLC Analysis of Vitamins, Fat Soluble (A and E) on Discovery® C18
HPLC Analysis of Vitamins, Fat Soluble (A and E) on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 30 C
mobile phase [A] methanol: [B] water (95:5, A:B)
flow rate 2 ml/min
injection 10 µl
detector UV, 290 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Food and Beverages
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC