Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Vitamins, Fat Soluble (A and E), on Discovery® C8
HPLC Analysis of Vitamins, Fat Soluble (A and E), on Discovery® C8
Conditions
column Discovery C8, 15 cm x 4.6 mm I.D., 5 µm particles (59353-U)
mobile phase [A] acetonitrile: [B] water (90:10 v/v, A:B)
flow rate 2 ml/min
injection 10 µl
detector UV, 290 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Food and Beverages
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC