HPLC Analysis of Vitamins, Water Soluble on Discovery® C18
HPLC Analysis of Vitamins, Water Soluble on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. ambient
mobile phase [A] 50 mM potassium phosphate, dibasic, pH 4.5 to methanol: [B] 50mM potassium phosphate, dibasic, pH 4.0 (30:70, A:B) in 8 min
flow rate 1 ml/min
injection 1 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Vitamins and Natural products
Featured Industry Food and Beverages
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC