Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Vitamins, Water Soluble, on Discovery® RP-Amide C16
HPLC Analysis of Vitamins, Water Soluble, on Discovery® RP-Amide C16
Conditions
column Discovery RP-Amide C16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 35 C
mobile phase [A] methanol; [B] 50 mM potassium phosphote, monobasic, pH 4.5 (30:70, A:B)
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Vitamins and Natural products, Anticonvulsant/antiepiletic/antiseizure
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC