HPLC Analysis of Vitamins, Water Soluble, on Ascentis® RP-Amide
HPLC Analysis of Vitamins, Water Soluble, on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 °C
mobile phase 25 mM potassium phosphate, dibasic (pH 3.5 with phosphoric acid)
flow rate 1.0 mL/min
sample as indicated in 25 mM potassium phosphate, dibasic (pH 3.5 with phosphoric acid)
injection 10 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Vitamins and Natural products, Anticonvulsant/antiepiletic/antiseizure
Featured Industry Vitamins, Nutraceuticals, and Natural Products