HPLC Analysis of Warfarin™ Anticoagulant on Ascentis® C18
HPLC Analysis of Warfarin™ Anticoagulant on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 C
mobile phase [A] 20 mM phosphoric acid (pH 2.0, unadjusted): [B] methanol; (30:70, A:B)
flow rate 1 ml/min
sample 50 µg/mL each in mobile phase
injection 10 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticoagulants
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, Warfarin is a trademark of Wisconsin Alumni Research Foundation