HPLC Analysis of Warfarin™ Enantiomers on Astec® CYCLOBOND® I 2000 (UV at 254 nm)
HPLC Analysis of Warfarin™ Enantiomers on Astec® CYCLOBOND® I 2000 (UV at 254 nm)
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 23 °C
mobile phase [A] acetonitrile: [B] acetic acid: [C] triethylamine; (100:0.3:0.25, A:B:C)
flow rate 1 mL/min
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, Chiral Applications, Anticoagulants, Clinical, Forensic & Toxicology, anticoagulants, Therapeutic Drug Monitoring
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC