HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 °C
mobile phase [A] 5 mM ammonium acetate, pH 4.1; [B] acetonitrile; (70:30, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Chiral Applications, Clinical, Clinical, Forensic & Toxicology, anticoagulants, Anticoagulants, Therapeutic Drug Monitoring
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC