HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 °C
mobile phase [A] 10 mM ammonium acetate, pH 4.1; [B] acetonitrile; (70:30, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Chiral Applications, Clinical, anticoagulants, Clinical, Forensic & Toxicology, Anticoagulants, Pharmaceutical (small molecule), Anticoagulants/Antiplatelet Drugs, All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC