HPLC Analysis of Warfarin™ Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Warfarin™ Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 °C
mobile phase [A] methanol; [B] acetic acid; [C] TEA; (100:0.2:0.1, A:B:C)
flow rate 0.5 mL/min
sample 2 mg/mL in mobile phase
injection 2 µL
detector UV, 278 nm
Description
Categories Analytical Chromatography, Chiral Applications, Clinical, anticoagulants, Anticoagulants, Clinical, Forensic & Toxicology, Therapeutic Drug Monitoring
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, Astec is a registered trademark of Sigma-Aldrich Co. LLC