HPLC Analysis of Warfarin™ Enantiomers on Astec® CYCLOBOND® I 2000 (UV at 278 nm)
HPLC Analysis of Warfarin™ Enantiomers on Astec® CYCLOBOND® I 2000 (UV at 278 nm)
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] acetic acid; [C] TEA; (100:0.3:0.2, A:B:C)
flow rate 1 mL/min
sample 2 mg/mL in mobile phase
injection 2 µL
detector UV, 278 nm
Description
Categories Analytical Chromatography, Anticoagulants
Featured Industry Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC