HPLC Analysis of White-Faced Hornet Venom on Mono S® HR5/5
HPLC Analysis of White-Faced Hornet Venom on Mono S® HR5/5
Conditions
column Mono S HR5/5, 10 µm particles (54808)
mobile phase [A] 50 mM BICINE, pH 8.4; [B] A + 0.35 M sodium chloride
flow rate 1 ml/min
injection 7 mg venom in mobile phase A
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Forensics and Toxicology
Life Science and Biopharma
Legal Information Mono S is a registered trademark of GE Healthcare