HPLC Analysis of Withania Standards on Discovery® C18
HPLC Analysis of Withania Standards on Discovery® C18
Conditions
column Discovery C18, 25 cm x 4.6 mm I.D., 5 µm particles (504971)
column temp. 27 C
mobile phase [A] 1 mM potassium phosphate, monobasic; [B] acetonitrile
gradient 5 to 45% B in 18 min; 45 to 80% B in 7 min; held at 80% B for 3 min
flow rate 1.5 ml/min
pressure 220 bar (3200 psi)
sample 20 µg/mL in water: acetonitrile (86:14)
injection 20 µl
detector UV, 227 nm
Description
Analysis Note This application demonstrates the suitability of Discovery C18 for the efficient separation of withania standards by the USP monograph.
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC