HPLC Analysis of Xanthines on Ascentis® C18
HPLC Analysis of Xanthines on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid); [B] methanol; (94:6, A:B)
flow rate 1 ml/min
sample 50 µg/mL each in mobile phase
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Bronchodilators
Featured Industry Life Science and Biopharma
Vitamins, Nutraceuticals, and Natural Products