HPLC Analysis of Xylo-oligosaccharide on Proteomix® SAX-NP5
HPLC Analysis of Xylo-oligosaccharide on Proteomix® SAX-NP5
Conditions
column Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 µm particles (Z777228)
mobile phase (A) 0.05% ammonium hydroxide in acetonitrile (B) 0.05% ammonium hydroxide in water
gradient 15-60% B (20 min)
flow rate 0.7 mL/min
pressure 1015 psi (70 bar)
temp. 35 °C
sample Xylose oligosaccharides
injection 10 µL
detector ELSD
Description
Categories Analytical Chromatography, Sweeteners
Featured Industry Food and Beverages
Legal Information Proteomix is a registered trademark of Sepax Technologies