Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Yohimbe Alkaloids on Ascentis® Express F5, HILIC Mode
HPLC Analysis of Yohimbe Alkaloids on Ascentis® Express F5, HILIC Mode
Conditions
column Ascentis Express F5, 3 cm x 3.0 mm I.D., 2.7 µm particles (53574-U)
column temp. 35 C
mobile phase 2 mM ammonium acetate in 10:90 water:acetonitrile, pH to 6.0 with acetic acid
flow rate 0.5 ml/min
pressure 1100 psi (76 bar)
sample 100 ug/mL each in methanol
injection 2 µl
detector UV, 275 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products