HPLC Analysis of Z-Drugs on Ascentis® Express C18 2.0 μm
HPLC Analysis of Z-Drugs on Ascentis® Express C18 2.0 μm
Conditions
column Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.0 µm particles (50819-U)
column temp. 30 C
mobile phase [A] 10 mM ammonium formate pH 3.0 with formic acid in 90:10, water:acetonitrile; [B] 10 mM ammonium formate pH 3.0 with formic acid in 10:90, water:acetonitrile;
gradient 0 to 100% B in 3.0 min; held at 100% B for 0.5 min
flow rate 0.6 ml/min
pressure 5405 psi (373 bar)
sample 10 µg/mL in 96:4, water:methanol
injection 0.5 µl
detector MS, ESI+, combined SIR
Description
Analysis Note An rapid method for the siµltaneous determination of the Z-drugs or sleep aids: zopiclone, zolpidem, and zaleplon is presented here. The need for greater analytical capacity and throughput for the analysis of sleep aid medicines (Z-drugs) in forensic toxicology laboratories can be met by the use of fast Ascentis Express 2.0 micron Fused Core UHPLC Columns.
Categories Analytical Chromatography, Anxiolytics, Sedatives
Featured Industry Clinical
Forensics and Toxicology