Hyper-Fast Separations on Ascentis® Express 2.7 μm at Half the Pressures of sub-2 μm Columns
Hyper-Fast Separations on Ascentis® Express 2.7 μm at Half the Pressures of sub-2 μm Columns
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 µm particles and sub-2 µm particle column, 5 cm x 2.1 mm I.D. (53823-U)
column temp. ambient
mobile phase [A] water; [B] acetonitrile; (55:45 or 54:46, A:B)
flow rate 0.2 ml/min
injection 1 µl
detector UV, 200 nm
Description
Categories Analytical Chromatography, Hormones