IC Separation of Chloride, Fluoride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 16
IC Separation of Chloride, Fluoride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 16
Conditions
column Metrosep A Supp 16, 15 cm x 4 mm I.D., 4.6 µm particles (Metrohm 6.1031.420)
column temp. 45 °C
mobile phase [A] 7.5 mM sodium carbonate; [B] 0.75 mM sodium hydroxide
flow rate 0.8 mL/min
sample chloride, fluoride (2 mg/L), nitrite (5 mg/L), bromide, nitrate, phosphate, sulfate (10 mg/L)
injection 10 µL
detector conductivity
Description
Categories Analytical Chromatography, Anions, Ion chromatography
Featured Industry Chemicals and Industrial Polymers
Environmental
Food and Beverages
General Analytical