Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
IC Separation of Chloride, Fluoride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 5
IC Separation of Chloride, Fluoride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 5
Conditions
column Metrosep A Supp 5, 15 cm x 4 mm I.D., 5 µm particles (Metrohm 6.1006.520)
column temp. 25 C
mobile phase [A] 3.2 mM sodium carbonate; [B] 1.0 mM sodium bicarbonate
flow rate 0.7 ml/min
sample chloride, fluoride (2 mg/L), nitrite (5 mg/L), bromide, nitrate, phosphate, sulfate (10 mg/L)
injection 20 µl
detector conductivity
Description
Categories Analytical Chromatography, Anions, Ion chromatography
Featured Industry Chemicals and Industrial Polymers
Environmental
Food and Beverages
General Analytical