IC Separation of Lithium, Sodium, Ammonium, Potassium, Calcium, and Magnesium Ions on Metrohm Metrosep C4
IC Separation of Lithium, Sodium, Ammonium, Potassium, Calcium, and Magnesium Ions on Metrohm Metrosep C4
Conditions
column Metrosep C 4, 15 cm x 4 mm I.D., 5 µm particles (Metrohm 6.1050.420)
column temp. 25 C
mobile phase [A] 1.7 mM nitric acid; [B] 0.7 mM dipicolinic acid
flow rate 0.9 ml/min
sample lithium (1 mg/L), sodium, ammonium (5 mg/L), potassium, calcium, magnesium (10 mg/L)
injection 10 µl
detector conductivity
Description
Categories Analytical Chromatography, Cations, Ion chromatography
Featured Industry Chemicals and Industrial Polymers
Environmental
Food and Beverages
General Analytical