LC/MS Analysis of Amphetamine Enantiomers on Astec CHIROBIOTIC® V2 in Urine after Solid Phase Extraction (SPE) using Supel™-Select SCX
LC/MS Analysis of Amphetamine Enantiomers on Astec CHIROBIOTIC® V2 in Urine after Solid Phase Extraction (SPE) using Supel™-Select SCX
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix 1 mL of synthetic urine spiked with 50 ng/mL of each enantiomer
SPE well plate Supel-Select SCX, 96-well plate, 30 mg/well (575664-U)
condition 1 mL 1% formic acid in acetonitrile followed by 1 mL water
sample addition 1 mL spiked urine sample
washing 2 mL water followed by 1 mL 5 mM ammonium phosphate dibasic in 50% methanol followed by 1 mL 25% v/v methanol in water
elution 1 mL 10% w/v ammonium hydroxide in acetonitrile
eluate post-treatment evaporate to dryness under nitrogen at 40 °C, reconstitute in 1 mL mobile phase
column Astec CHIROBIOTIC V2, 15 cm x 4.6 mm I.D., 5 µm particles (15023AST)
column temp. 20 °C
mobile phase [A] methanol; [B] water; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.1:0.02, A:B:C:D)
flow rate 1 mL/min
pressure 1220 psi (84 bar)
injection 2 µL
detector MS, ESI(+),SIR m/z 150.1
Description
Analysis Note Shown here is the chiral separation of amphetamine enantiomers under MS-compatible conditions on Astec CHIROBIOTIC V2 after extraction from urine using Supel-Select SCX SPE 96-well plates. The highest grade mobile phase solvents and additives were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Categories Analytical Chromatography, Forensics and Toxicology, enantiomers, drugs of abuse, Drugs of Abuse, Enantiomers
Featured Industry Forensics and Toxicology
Legal Information Supel is a trademark of Sigma-Aldrich Co. LLC, CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS, application for SPE