LC/MS Analysis of Amphetamines on Ascentis® Express HILIC
LC/MS Analysis of Amphetamines on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 3 mm I.D., 2.7 µm particles (53970-U)
column temp. 35 °C
mobile phase [A] 1 mM ammonium acetate in [B] 10:90 (v/v) water:acetonitrile
flow rate 0.6 mL/min
pressure 1189 psi (82 bar)
sample 10 mg/L each in acetonitrile
injection 2 µL
detector ESI(+), full scan, m/z 110-200
Description
Categories Analytical Chromatography, Forensics and Toxicology, drugs of abuse, Drugs of Abuse
Featured Industry Forensics and Toxicology