Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Amphetamines on Ascentis® Express F5
LC/MS Analysis of Amphetamines on Ascentis® Express F5
Conditions
column Ascentis Express F5, 5 cm x 3.0 mm, 2.7 µm particles (53576-U)
column temp. 25 C
mobile phase 5 mM Ammonium acetate in 95:5, acetonitrile:water
flow rate 0.6 ml/min
pressure 2540 psi (175 bar)
sample 500 ng/mL acetonitrile
injection 1 µl
detector MS ESI+; SIR
Description
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Life Science and Biopharma
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS