Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Ancymidol Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
LC/MS Analysis of Ancymidol Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
column temp. 35 C
mobile phase [A] 5 mM ammonium acetate, pH 6.0; [B] acetonitrile; (80:20, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 5 µl
detector MS (m/z = 257)
Description
Categories Analytical Chromatography, Chiral Applications, Environmental, Pesticides & Herbicides, All Environmental Applications, Herbicides
Featured Industry Agriculture
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC