LC/MS Analysis of Antidepressants From Urine Using an Ascentis® Express C18 Column
LC/MS Analysis of Antidepressants From Urine Using an Ascentis® Express C18 Column
Conditions
column Ascentis Express C18, 10cm x 2.1mm, 2.7 µm particles (53823-U)
column Ascentis Express C18, 10cm x 2.1mm, 2.0 µm particles (50813-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium acetate in 0.1% formic acid; [B] 0.1% formic acid in acetonitrile
gradient 10% B held  for 1 min; to 55% B in 10 min
flow rate 0.4 mL/min
pressure 8280 psi (570 bar)
injection 1 µL
detector MS, ESI(+), MRM, m/z 247.138/71.958,  278.168/58.025, 372.054/148.011,   442.114/155.035,  298.014/43.96, 305.968/158.873, 325.128/108.95
Description
Analysis Note Seven modern antidepressants were analyzed from urine after a dilute and inject method using reversed-phase chromatography and mass spectrometric detection.
Categories Analytical Chromatography, Antidepressants
Featured Industry Pharmaceutical (small molecule)
General description Sample preparation: Prepare a stock solution with 100 ng/mL of compounds in synthetic urine (Cerilliant, part number S-020, lot 72112). Dilute this solution with 95:5 (5mM ammonium acetate in 0.1% formic acid in water: 0.1% formic acid in acetonitrile) to give a final concentration of 50 ng/mL. Filter through a Millex LH filter. Inject. Also prepared blank in same way.
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS