LC/MS Analysis of Azithromycin on Titan™ C18
LC/MS Analysis of Azithromycin on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 50 C
mobile phase 20 mM ammonium acetate pH unadjusted in 40:60, water:methanol
flow rate 0.4 ml/min
pressure 10091 psi (696 bar)
sample azithromycin 500 ng/mL in 40:60, water:methanol
injection 2 µl
detector MS, SIR, m/z 749.5
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS