LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide
LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 5.0 with acetic acid): [B] acetonitrile; (20:80, A:B)
flow rate 1 mL/min, split to the MS
sample 1 µg/mL in water:acetonitrile (1:1)
injection 5 µL
detector MS ESI (+), SIR Mode
Description
Categories Analytical Chromatography, Forensics and Toxicology, Pharmaceutical Analysis, Analgesics, drugs of abuse, Anti-inflammatories
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)